Learn More
Hettich Lab Technology™ Filtre
Filter Cards are used for collecting cyto centrifuge samples and come in sizes to accommodate a variety of chambers and carriers.
Marque: Hettich Lab Technology™ 1675
Code nomenclature Nacres: NB.82
Informations supplémentaires : Poids : 0.90000kg
Description
- For cyto centrifuge systems and cell evaluations
- Suitable for a variety of cyto chambers and slide carriers
Spécification
Filtre | |
1 mL |