Learn More
Fistreem International™ Pre-Treat Feed Accessory Kit
Suitable pre-treatment feed kit for the Calypso Water Still. Fistreem International™ Pre-Treat Feed Accessory Kit allows you to connect a suitable pre-treated water supply ( deionized, reverse osmosis or already distilled water ) to your still.
Marque: Fistreem International™ WCA901.SVA.7
Code nomenclature Nacres: JA.13
Informations supplémentaires : Poids : 5.00000kg
Description
- Allows you to connect a suitable pre-treated water supply to your still.
- In the pre-treated mode, the already purified water is fed directly to the boiler and tap water is fed to the condenser coil for cooling purposes only.
Spécification
Calypso Water Still | |
Pre-treat Feed Accessory Kit |