Learn More
Fc gamma RIIB/CD32b Antibody (2E10), Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals H00002213-M01-100ug
Code nomenclature Nacres: NA.46
Code produit. 18314468
  • 613.00€ / 100µg
Description
Fc gamma RIIB/CD32b Monoclonal antibody specifically detects Fc gamma RIIB/CD32b in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Fc gamma RIIB/CD32b
Monoclonal
Unconjugated
In 1x PBS, pH 7.4
CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b
FCGR2B (AAH31992, 45 a.a. ~ 154 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AAPPKAVLKLEPQWINVLQEDSVTLTCRGTHSPESDSIQWFHNGNLIPTHTQPSYRFKANNNDSGEYTCQTGQTSLSDPVHLTVLSEWLVLQTPHLEFQEGETIVVRCHS
100 μg
Immunology
2213
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
2E10
Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
AAH31992
Mouse
IgG purified
RUO
Primary
Human
Purified