missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETS2 Mouse anti-Human, Clone: OTI2A3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marque: Origene Technologies CF505391
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes.Spécification
ETS2 | |
Monoclonal | |
Unconjugated | |
ETS2 | |
ETS2IT1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2114 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2A3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P15036 | |
ETS2 | |
Human recombit protein fragment corresponding to amino acids 90-349 of human ETS2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |