Learn More
Ethynyl Estradiol, MedChemExpress
MedChemExpress Ethynyl Estradiol (17α-Ethynylestradiol;Ethynylestradiol) is an orally bio-active estrogen used in almost all modern formulations of combined oral contraceptive pills.
Marque: Medchem Express HY-B0216
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Ethynyl Estradiol (17α-EthynylestradiolSpécification
Ethynyl Estradiol | |
White | |
C20H24O2 | |
17α-Ethynylestradiol Ethynylestradiol | |
OC1=CC=C2C(CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4(C#C)O)=C1 | |
296.4 | |
Research | |
Cancer-programmed cell death |
57-63-6 | |
Research | |
500 mg | |
DMSO : 250 mg/mL (843.45 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) | |
296.4 | |
99.28% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only
Spot an opportunity for improvement?Share a Content Correction