Learn More
Éthyl 1-Boc-pipérazine-3-carboxylate, 97 %, Thermo Scientific Chemicals
Code produit. 15459678
Click to view available options
Quantité:
250 mg
1 g
5 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 183742-29-2
Formule moléculaire C12H22N2O4
Poids moléculaire (g/mol) 258.318
Numéro MDL MFCD13172266
Clé InChI IQKOVLZJPVVLOZ-UHFFFAOYSA-N
Synonyme 1-tert-butyl 3-ethyl piperazine-1,3-dicarboxylate, 1-boc-3-piperazinecarboxylic acid ethyl ester, 1-n-boc-3-piperazinecarboxylic acid ethyl ester, 1-tert-butoxycarbonyl-3-ethoxycarbonylpiperazine, 1-tert-butyl3-ethylpiperazine-1,3-dicarboxylate, 1-tert-butyl-3-ethylpiperazine-1,3-dicarboxylate, ethyl 4-tert-butoxycarbonyl-2-piperazinecarboxylate, 1-1,1-dimethylethyl 3-ethyl 1,3-piperazinedicarboxylate, 1-tert-butoxycarbonylpiperazine-3-carboxylic acid ethyl ester, ethyl 4-1,1-dimethylethoxycarbonyl-2-piperazinecarboxylate
CID PubChem 11402671
Nom IUPAC 1-O-tert-butyl 3-O-éthyl pipérazine-1,3-dicarboxylate
SMILES CCOC(=O)C1CN(CCN1)C(=O)OC(C)(C)C
Spécification
Nom chimique ou matériau Ethyl 1-Boc-piperazine-3-carboxylate
Quantité 250 mg
Poids de la formule 258.31
Pourcentage de pureté 97%
Forme physique Pleine