Learn More
Enoblituzumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Marque: Invitrogen MA542090
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Enoblituzumab is a biosimilar that targets CD276.Spécification
Enoblituzumab | |
Recombinant Monoclonal | |
Unconjugated | |
MG-A271 | |
Human CD276/B7-H3 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
2.04 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 κ |