missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enasidenib, MedChemExpress
missing translation for 'orderingAttributeHoverText'
Quantité:
5 mg
10 mg
50 mg
100 mg
Description
Enasidenib is an oral, potent, reversible, selective inhibitor of the IDH2 mutant enzymes, with IC50s of 100 and 400 nM against IDH2R140Q and IDH2R172K, respectively.
Specifications
Specifications
| Nom chimique ou matériau | Enasidenib |
| CAS | 1446502-11-9 |
| Couleur | Blanc |
| Notes de qualité de la pureté | Research |
| Formule moléculaire | C19H17F6N7O |
| Quantité | 5 mg |
| Synonyme | AG-221 |
| Informations sur la solubilité | DMSO : 50 mg/mL (105.62 mM; Need ultrasonic) |
| SMILES | CC(O)(C)CNC1=NC(C2=NC(C(F)(F)F)=CC=C2)=NC(NC3=CC(C(F)(F)F)=NC=C3)=N1 |
| Poids moléculaire (g/mol) | 473.38 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction