missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGTA, MP Biomedicals
Click to view available options
Quantité:
50 g
Spécification
Spécification
Nom chimique ou matériau | EGTA |
CAS | 67-42-5 |
Formule moléculaire | C14H24N2O10 |
Quantité | 50 g |
Synonyme | egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn |
Clé InChI | DEFVIWRASFVYLL-UHFFFAOYSA-N |
SMILES | C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O |
Nom IUPAC | Acide 2-[2-[2-[2-[bis(carboxyméthyl)amino]éthoxy]éthoxy]éthyle-(carboxyméthyl)amino]acétique |
Poids moléculaire (g/mol) | 380.35 |
CID PubChem | 6207 |
Afficher plus |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'