missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFTUD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10208-1-AP-150UL
This item is not returnable.
View return policy
Description
10208-1-AP is a rabbit polyclonal antibody raised against the N-terminal of human EFTUD2.
EFTUD2 is component of the U5 snRNP complex required for pre-mRNA splicing.Specifications
EFTUD2 | |
Polyclonal | |
Unconjugated | |
EFTUD2 | |
EFTUD2, hSNU114, KIAA0031, Snrp116, Snu114, SNU114 homolog, U5 116 kDa, U5 116KD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20624, 287739, 9343 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O08810, Q15029 | |
Eftud2 | |
EFTUD2 Fusion Protein Ag0279 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |