missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20107-1-AP-20UL
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
Catalyzes the GTP-dependent ribosomal translocation step during translation elongation. During this step, the ribosome changes from the pre-translocational (PRE) to the post-translocational (POST) state as the newly formed A-site-bound peptidyl-tRNA and P-site-bound deacylated tRNA move to the P and E sites, respectively. Catalyzes the coordinated movement of the two tRNA molecules, the mRNA and conformational changes in the ribosome.Spécification
EEF2 | |
Polyclonal | |
Unconjugated | |
EEF2 | |
EEF 2, EEF2, EF 2, EF2, Elongation factor 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13629, 1938 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P13639, P58252 | |
EEF2 | |
EEF2 Fusion Protein Ag13826 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |