missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 45 mm Shelf Height

Eco AluRack for upright freezers is designed to hold microtiterplates.

Brand:  Tenak™ TE15482

 View more versions of this product

Product Code. 12488230

  • 115.00€ / 1 pièce
Expédition estimée: 06-05-2025
pour voir le stock disponible 
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • À utiliser dans des congélateurs verticaux
  • Les racks peuvent être utilisés avec une plaque par étagère ou les plaques peuvent être empilées
  • Fabrication en aluminium
Specifications

Specifications

4 x 5
Aluminium
Congélateur vertical
45 mm
Plaques de microtitration (20)
20
450 x 135 x 186 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.