Learn More
STEMCELL Technologies EasySep™ Non-Human Primate T Cell Isolation Kit, with RoboSep™ Buffer and Filter Tips
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
Marque: STEMCELL Technologies 19581RF
Informations supplémentaires : Poids : 0.75220kg
Description
- The EasySep™ Non-Human Primate T Cell Isolation Kit is designed to isolate T cells from fresh peripheral blood mononuclear cells by immunomagnetic negative selection
- The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
- The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
- This kit has been verified for use with rhesus and cynomolgus macaques
Spécification
T Cells | |
Cell Isolation | |
1 x 109 cells |