Learn More
STEMCELL Technologies EasySep™ Non-Human Primate T Cell Isolation Kit, with RoboSep™ Buffer and Filter Tips
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
Marque:  STEMCELL Technologies 19581RF
Informations supplémentaires : Poids : 0.75220kg
Code produit. 17107931
  • 1657.50€ / 1 kit
Expédition estimée: 23-07-2024
pour voir le stock disponible 
Ajouter au panier
Description
Description
  • The EasySep™ Non-Human Primate T Cell Isolation Kit is designed to isolate T cells from fresh peripheral blood mononuclear cells by immunomagnetic negative selection
  • The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
  • The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
  • This kit has been verified for use with rhesus and cynomolgus macaques
Spécification
Spécification
T Cells
Cell Isolation
1 x 109 cells