Learn More
STEMCELL Technologies EasySep™ Mouse Pan-B Cell Isolation Kit
Immunomagnetic isolation of untouched mouse pan-B cells
Marque: STEMCELL Technologies 19844
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- The EasySep™ Mouse Pan-B Cell Isolation Kit is designed to isolate all B cells, including conventional B-2 B cells, B-1 B cells and plasma cells from single-cell suspensions of splenocytes or other tissues by negative selection
- Unwanted cells are targeted for removal with biotinylated antibodies directed against non pan-B cells (CD4, CD8, CD11c, CD49b, CD90.2, Ly-6C/G (Gr-1), TER119) and streptavidin-coated magnetic particles (RapidSpheres™ )
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
- This kit is compatible for use with cells from disease models where the malignant cells (B-CLL) express CD43 or CD11b
- For isolation of conventional B cells only, we recommend using the EasySep™ Mouse B Cell Isolation Kit (Catalog #19854)
Spécification
B Cells | |
Cell Isolation | |
1 x 109 cells |