Learn More
Berkshire Corporation EasyClean™ 360 Isolator Knitted Polyester Padded Covers
Tous les produits Berkshire Produits
Click to view available options
Couleur:
Blanc
Conditionnement:
200 pièces
Description
- Made from Berkshire's Choice™ SuperSorb 100% continuous filament knitted polyester material
- Laundered and packaged in an ISO Class 4 cleanroom for superior fiber, particle and residue performance
- Foam-free cushioned design ensures even surface coverage with reduced residues and contaminants
- Easy installation and secure fit on the mop head
- Recommended for use with the EasyClean™ 360 Knitted Polyester Elastic Covers
- Compatible for use in ISO Class 3/EU Grade A environments after sterilizing
Spécification
Spécification
| Type de produit | Mop Cover |
| Code couleur | White |
| Couleur | Blanc |
| Stérilité | Non stérile |
| Autoclavable | Autoclavable |
| Jetable | Usage unique |
| Color-coded | No |
| Matériau | Polyester |