Learn More
DUSP16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14237-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a mitogen-activated protein kinase phosphatase that is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. The encoded protein specifically regulates the c-Jun amino-terminal kinase and extracellular signal-regulated kinase pathways.Spécification
DUSP16 | |
Polyclonal | |
Unconjugated | |
DUSP16 | |
DUSP16, KIAA1700, MAP kinase phosphatase 7, MKP 7, MKP7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
297682, 70686, 80824 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BY84 | |
Dusp16 | |
DUSP16 Fusion Protein Ag5479 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |