Learn More
LABPLAS Dry Polyurethane Sani-Stick
Click to view available options
Comprend:
Aucun
Gants
Conditionnement:
100 pièces
Description
- Sani-Stick
- Dry polyurethane sponge with handle
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Spécification
Spécification
| Type de produit | Hygiene Sponge Sampling Kit |
| Stérilité | Stérile |
| Comprend | Aucun |