missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOPA decarboxylase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10166-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
DOPA Decarboxylase (aromatic L-amino acid decarboxylase, AADC; DDC) catalyzes the second reaction in the biosynthesis of catecholamines and serotonin (Waymire and Haycock, 2002; Berry et al., 1996; Haycock et al., 2003). It is also involved in the biosynthesis of trace amines. DDC antibodies can therefore be used as markers for dopaminergic, noradrenergic and serotonergic neurons in a variety of applications including depression, schizophrenia, Parkinson′s disease and drug abuse (Kish et al., 2001; Zhu et al., 2000; Zhu et al., 1999).Spécification
DOPA decarboxylase | |
Polyclonal | |
Unconjugated | |
DDC | |
AADC, DDC, DOPA decarboxylase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13195, 1644, 24311 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88533, P14173, P20711 | |
DDC | |
DOPA decarboxylase Fusion Protein Ag0219 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu