Learn More
Invitrogen™ Donkey anti-Rabbit IgG (H+L) Secondary Antibody, Qdot™ 625
Donkey Polyclonal Secondary Antibody
Marque:  Invitrogen™ Q22086
Code nomenclature Nacres: NA.46
Code produit. 12313643
Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
Voir les produits de remplacement »
Description
Description
Qdot nanocrystals are composed of semi-conductor material to generate a fluorescent particle which is exceptionally bright and does not photobleach. Qdot nanocrystals paired with the correct optical filters are as much as 50 times brighter than traditional organic dyes.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Rabbit IgG (H+L)
Polyclonal
Qdot 625
Donkey
Purified
RUO
Rabbit
Secondary Antibody
IgG
Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry
1 μM
0.05M borate with 1M betaine and 0.05% sodium azide; pH 8.3
Gamma Immunoglobins Heavy and Light chains.
100 μL
Secondary
4°C, store in dark