missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Mouse IgG (H+L) Secondary Antibody, DyLight 680, Novus Biologicals™
Donkey Polyclonal Antibody has been used in 1 publication
Marque: Novus Biologicals NBP1-72936
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.Spécification
IgG (H+L) | |
Polyclonal | |
DyLight 680 | |
Donkey | |
Affinity Purified | |
RUO | |
Secondary | |
Reconstitute with 100 ul deionized water (or equivalent). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
LYOPH | |
Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:5000, Fluorophore-linked immunosorbent assay 1:20000 | |
Mouse IgG whole molecule | |
0.1 mg | |
Adaptive Immunity, Immunology | |
This antibody will react with heavy chains of Mouse IgG and with light chains of most Mouse immunoglobulins. | |
Mouse | |
IgG |
For Research Use Only