Learn More
Donkey anti-Mouse IgG (H+L) Secondary Antibody, DyLight 650 (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Marque: Novus Biologicals NB120-7056C
286.40 EUR valable jusqu'au 2025-03-28
Utilisez le code promo "25306" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spécification
IgG (H+L) | |
Polyclonal | |
50mM Sodium Borate | |
Donkey anti-Mouse IgG (H+L) Secondary Antibody (Pre-adsorbed) was produced by repeated immunization with Mouse IgG whole molecule in donkey. | |
0.5 mL | |
Adaptive Immunity, Immunology | |
Mouse | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Fluorophore-Linked Immunosorbent Assay | |
DyLight 650 | |
Donkey | |
Multi-step | |
RUO | |
Secondary | |
Store at 4°C in the dark. |