Learn More
DNA Ligase I Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque:  Novus Biologicals NBP2-97831APC
Code nomenclature Nacres: NA.46
Code produit. 18635662
  • 407.00€ / 0.10mL
Description
DNA Ligase I Polyclonal antibody specifically detects DNA Ligase I in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Produced in rabbits immunized with E. coli-derived Human DNA Ligase I fragment.
Antigen and protein A Affinity-purified