Learn More
Interscience™ Dispensing Assembly for Dilutors
Set includes parts required for dispensing with dilutors.
Marque: Interscience™ 513043
Code nomenclature Nacres: NB.05
Informations supplémentaires : Poids : 0.20000kg
Description
Includes:
- Set of silicone tubings with inner diameter of 6.4 mm
- GL45 screw cap
- Tube weight
- Nozzle
- Filter
Spécification
Dispensing Tubing | |
Autoclavable | |
Adding Dispensing Assembly to Dilutor | |
DiluFlow, DiluFlow Pro, DiluFlow Elite |