Learn More
TFA Dostmann Digital Fridge/Freezer Thermometer
The digital fridge-freezer thermometer will show a symbol on the display depending on the temperature. The stored highest and lowest temperatures are permanently displayed. This makes it easy to determine if the cooling has been interrupted. The thermometer is EN 13485 compliant.
Marque:  TFA Dostmann 30.1042
Code produit. 15556385
  • 14.35€ / 1 pièce
Expédition estimée: 11-04-2025
pour voir le stock disponible 
Découvrez plus d'offres spéciales
Ajouter au panier
Description
Description
This useful thermometer shows you at a glance, whether vegetables, cheese, meat and other food are properly cooled. So food getting spoiled prematurely becomes a thing of the past. A freezer or fridge symbol will be shown on the display depending on the temperature. The stored highest and lowest temperatures are permanently displayed. This makes it easy to determine if the refrigerator door has been open for a long time or if a power cut has interrupted cooling. The thermometer has a fold-out stand. With the self-adhesive frame, you can attach the thermometer on a variety of surfaces, even behind the glass in the vegetable compartment or the refrigerated display case. The thermometer is EN 13485 compliant and ideally suited for professional use in industry, commerce, trade and laboratory.
  • For temperature measurement in the fridge and freezer
  • Permanent display of max. and min. values
  • For mounting on surfaces or behind glass
  • With freezer or fridge symbol
  • Also for professional use, compliant with EN13485
Spécification
Spécification
Plastic
Oui
LCD
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats
Promotions
Promotions
Correction du contenu d'un produit
Nom du produit
TFA Dostmann Digital Fridge/Freezer Thermometer > Affichage : LCD