missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmin Mouse, Clone: DES/1711, enQuireBio™
Mouse Monoclonal Antibody
Marque: N/A 1674-MSM1-P1
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Spécification
Desmin | |
Cytoplasmic | |
DES/1711 | |
Unconjugated | |
NP_001918 | |
Mouse | |
52kDa | |
Protein A/G Purified | |
RUO | |
1674 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
DES | |
Recombinant human full-length desmin protein | |
SJRH30 Cells. Uterus or Leiomyosarcoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |