Learn More
Invitrogen™ Desmin Monoclonal Antibody (D9)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA106401
Description
MA1-06401 detects desmin in human, mouse, rat, rabbits, pig, and chicken samples. MA1-06401 has sucessfully been used in Western blotting, and immunohistochemistry. By Western blotting, it detects a ~53 kDa protein representing desmin. The MA1-06401 immunogen is desmin isolated from human leiomyoma.
Desmin (approximately 53 kDa) exhibits a high degree of tissue specificity, its expression being predominantly confined to all types of muscle cells (cardiac, skeletal and smooth muscle). Regulation of desmin expression is stage and tissue-specific, since it is induced during terminal development of, for example, skeletal muscle cell differentiation. In skeletal en cardiac muscle cells desmin is localized in the Z-disk region and at the intercalated disk. The expression pattern of desmin in smooth muscle is much more heterogenous. Coexpression of vimentin and desmin has been observed in tumors derived from muscle tissue, i.e. rhabdomyosarcomas and leiomyosarcomas. Furthermore, during myocard dysfunction dramatic changes in the distribution of desmin have been observed.
Spécification
| Desmin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P02540, P02542, P17661, P31001, P48675 | |
| DES | |
| Desmin isolated from human leiomyoma. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Pig, Chicken | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| D9 | |
| Unconjugated | |
| DES | |
| CMD1I; CSM1; CSM2; DES; Desmin; desmin-like protein; FLJ12025; FLJ39719; FLJ41013; FLJ41793; I79_018932; intermediate filament protein; LGMD2R; MFM1; muscle-specific intermediate filament desmin; muscle-specific intermediate filament protein; mutant desmin p.K241E; SCPNK; similar to desmin | |
| Mouse | |
| Purified | |
| RUO | |
| 100328890, 13346, 1674, 395906, 396725, 64362 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.