Learn More
Abnova™ Deoxyribonuclease I (Bovine), Molecular Biology Grade
Code produit. 16171830
Click to view available options
Quantité:
2 500 U
Conditionnement:
2500 unités
Code produit. 16171830
Marque: Abnova™ P5308.2500Units
Used for Enzyme
#REF!
Spécification
Numéro d’adhésion P00639
À utiliser avec (application) Enzyme Activity
Identification génétique (Entrez) 282217
Poids moléculaire 31.3kDa
Nom Deoxyribonuclease I (Bovine), Molecular Biology Grade
Méthode de préparation Native protein chromatographically purified from Bovine Pancreas
Quantité 2 500 U
Conditions de stockage Store at 4°C on dry atmosphere. Each vial contains approximately 2 mg glycine and 2 umoles calcium per 10,000 units of DNase I. Reconstituted the entire vial in 5 ml provides the equivalent of a 1 mg/ml solution.
État réglementaire RUO
Nom usuel DNASE1
Symbole de gène(s) DNASE1
Dosages Deoxyribonuclease I Assay; Nick translation; DNA mapping; Isolation of nuclear RNA and protein; Plasmid construction; RNA polymerase synthesis of RNA probes and RT-PCR.
Activité biologique ≥2,000 Kunitz units per mg dry weight. One Unit causes an increase in absorbance at 260nm of 0.001 per minute per ml at 25°C when acting upon highly polymerized DNA at pH 5.0.
Espèces Bovine
Marqueur de protéine None
Système d’expression Native protein chromatographically purified from Bovine Pancreas
Forme Lyophilized