Learn More
DDX21 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NB100-1718MFV450
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
DDX21 Polyclonal antibody specifically detects DDX21 in Human, Mouse, Zebrafish samples. It is validated for Flow Cytometry
Spécification
| DDX21 | |
| Flow Cytometry | |
| Flow Cytometry | |
| Store at 4C in the dark. |