Learn More
Tocris Bioscience™ Cy5B-trisulfo, NHS
Restricted conformation cyanine dye with high quantum yield and long fluorescence lifetime for advanced microscopy applications
Marque:  Tocris Bioscience™ 7394/300U
Code nomenclature Nacres: NA.22
Informations supplémentaires : Poids : 0.00970kg
Code produit. 18064442
  • 385.12€ / 300µg
Expédition estimée: 28-08-2024
pour voir le stock disponible 
Description
Description
Key information: Cy5B-trisulfo, NHS is a far-red fluorescent dye; supplied with an NHS ester reactive group for the labeling of primary amines. Particularly useful for smFRET experiments to reduce blinking and bleaching artefacts and maintain correct mean FRET efficiencies. Application: FRET, smFRET, FLIM, TIRF, and super resolution microscopy (SRM) techniques, including dSTORM. Provides enhanced spatial resolution in dSTORM experiments. Does not provide efficient blinking in conventional high-thiol dSTORM buffers; instead, sodium borohydride (NaBH4) addition is required following which SmLM experiments can be performed in PBS. Properties and Photophysical Data: Excitation and emission maxima (λ) are 669 nm and 685 nm, respectively; extinction coefficient = 241,000 M-1cm-1; quantum yield = 0.4; fluorescence lifetime = 1.85 ns. We suggest making up a 10 mM stock solution of this product in anhydrous DMSO. For more information on how Cy5B-trisulfo, NHS may be used, see our protocol: Conjugation Protocol for Amine Reactive Dyes .
Spécification
Spécification
300 μg
Fluorescent Dyes
C47H52N4Na2O15S3