Learn More
Thermo Scientific™ CultiMaxx™ Shelving System for G-Rex 500M-CS Bioreactors
Code produit. p-200008781 Tous les produits Thermo Scientific Produits
Click to view available options
Nbre d’étagères:
7
Matériau:
Acier inoxydable
Conditionnement:
1 kit
Code produit. 17988084
Marque: Thermo Scientific™ 332395G08
Il en reste null
Ajouter au panier
Ajouter au panier
CultiMaxx™ Shelving Systems to support G-Rex 500M-CS Bioreactors
Maximize the capacity, increase throughput and optimize cell growth in your lab using the CultiMaxx shelving system in a range of large capacity CO2 Incubators.
Maximize incubator space  
  • These shelving systems are designed to support scale-out of the G-Rex 500M-CS* in cell therapy production.
  • With an in-depth review of the shelving design, this new system provides up to a 150% increase in capacity for the G-Rex, or another way to look at it: 2 1/2 incubators in 1.
 Easy installation
  • Shelving systems are retrofittable and can be self-installed.
*G-Rex is a registered trademark of Wilson Wolf Corporation. G-Rex bioreactors must be ordered from Wilson Wolf Corporation.
TRUSTED_SUSTAINABILITY
Spécification
Description Shelving system to support G-Rex™ 500M for Reach-In 821L CO2 Incubator
À utiliser avec (équipement) Reach-In 821L CO2 Incubator
Matériau Acier inoxydable
Nbre d’étagères 7
Type Shelf
Comprend 7 levels of shelving