missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ COX6B1 (Human) Recombinant Protein
Human COX6B1 full-length ORF ( AAH01015, 1 a.a. - 86 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00001340-P01.25ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Sequence: MAEDMETKIKNYKTAPFDSRFPNQNQTRNCWQNYLDFHRCQKAMTAKGGDISVCEWYQRVYQSLCPTSWVTDWDEQRAEGTFPGKI
Spécification
AAH01015 | |
cytochrome c oxidase subunit Vib polypeptide 1 (ubiquitous) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COX6B1 | |
GST |
1340 | |
Wheat germ expression system | |
25 μg | |
COX6B, COXG | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |