Learn More
Fil de nickel et cuivre, 0,5 mm (0,02 po) de diamètre, Thermo Scientific Chemicals
41.81€ - 257.28€
Spécification
Nom chimique ou matériau | Copper Nickel wire |
---|---|
Note relative au nom | 0.5mm (0.02 in.) dia. |
Moy. Mol. Poids ou mol. Plage de poids | Cu:Ni 55:45 wt% |
Numéro MDL | MFCD00801098 |
Informations sur la solubilité | Insoluble in water. |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
11399378
|
Thermo Scientific Alfa Aesar
043272.G9 |
25 m |
41.81€
25m |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
11389378
|
Thermo Scientific Alfa Aesar
043272.H4 |
100 m |
108.54€
100m |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
11309388
|
Thermo Scientific Alfa Aesar
043272.H9 |
500 m |
257.28€
500m |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Copper Nickel wire is employed for sea water condenser systems and in desalination plants, as well as pipe work in chemical plants. It is being increasingly employed for brake and hydraulic suspension systems and cooling systems in cars and commercial vehicles. It is commonly used for the manufacture of coins, medals and other semi valuable applications. It is also used in cooling circuits, ammunition, sea water corrosion-resistant assemblies and condenser tubes. The good resistance to biofouling and sea water corrosion resistance of the copper nickel alloys have lead to its use in applications such as cladding for ships' hulls, legs of oil rig platforms and sea water intake screens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Copper Nickel wire | |
Cu:Ni 55:45 wt% | |
Insoluble in water. | |
Câble |
0.5mm (0.02 in.) dia. | |
MFCD00801098 | |
≈1.75g/m |
Sécurité et traitement
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350-H372
missing translation for 'tsca' : Oui
Recommended Storage : Température ambiante
RUO – Research Use Only