Learn More
Fistreem International™ Constant Level Device
Replacement constant level device for the Calypso or Cyclon Ultrapure Water Still. Fistreem International™ Constant Level Device allows cleaning solution to be simply introduced into the boiler with no dismantling.
Marque: Fistreem International™ WSC707.CLJ.9
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Borosilicate glass construction.
- Suitable for Calypso or Cyclon Ultrapure Water Still.
Spécification
5 kg | |
Calypso or Cyclon Ultrapure Water Still | |
Constant Level Device |