missing translation for 'onlineSavingsMsg'
Learn More

Concanavalin A, Thermo Scientific Chemicals

Code produit. 16348158
Click to view available options
Quantité:
250 mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

A plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts

Concanavalin A is a plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Concanavalin A is a plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts.

Solubility
Slightly soluble in water and phosfate buffered saline.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 11028-71-0
Numéro MDL MFCD00071069
Synonyme ConA

Spécification

Nom chimique ou matériau Concanavalin A
Type Concanavalin A
Forme physique Lyophilized Powder
Merck Index 14,2495
Informations sur la solubilité Slightly soluble in water and phosfate buffered saline.
Couleur Blanc
Quantité 250 mg

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'