missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement factor B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10170-1-AP-20UL
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody raised against 573-761aa of human complement factor B can recognize complement factor B and Bb fragment.
This gene encodes complement factor B, a component of the alternative pathway of complement activation. Factor B circulates in the blood as a single chain polypeptide. Upon activation of the alternative pathway, it is cleaved by complement factor D yielding the noncatalytic chain Ba and the catalytic subunit Bb. The active subunit Bb is a serine protease which associates with C3b to form the alternative pathway C3 convertase. Bb is involved in the proliferation of preactivated B lymphocytes, while Ba inhibits their proliferation. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. This cluster includes several genes involved in regulation of the immune reaction. Polymorphisms in this gene are associated with a reduced risk of age-related macular degeneration. The polyadenylation site of this gene is 421 bp from the 5′ end of the gene for complement component 2.Spécification
Complement factor B | |
Polyclonal | |
Unconjugated | |
CFB | |
BF, BFD, C3/C5 convertase, CFAB, CFB, Complement factor B, FB, FBI12, GBG, Glycine rich beta glycoprotein, H2 Bf, PBF2, Properdin factor B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
629 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P00751 | |
CFB | |
Complement factor B Fusion Protein Ag0226 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |