missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen II Antibody (5B2.5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 23 publications
Marque: Novus Biologicals NB600-844
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Collagen II Monoclonal specifically detects Collagen II in Human, Mouse, Rat, Chicken, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Spécification
| Collagen II | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 μg/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:2000, Immunohistochemistry-Frozen 1:10-1:500 | |
| Alpha-1 type II collagen, ANFH, AOM, cartilage collagen, chondrocalcin, COL11A3, Collagen 2, collagen alpha-1(II) chain, collagen II, alpha-1 polypeptide, collagen type 2, collagen, type II, alpha 1, collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphysealdysplasia, congenital), Collagen-2, MGC131516, SEDC | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Rat reactivity reported in scientific literature (PMID: 24349545). | |
| Store at 4C. Do not freeze. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 5B2.5 | |
| Unconjugated | |
| Q90W37 | |
| COL2A1 | |
| A purified preparation of lathyritic type II collagen from embryonic chicken sternum | |
| 0.1 mL | |
| Cytoskeleton Markers, Extracellular Matrix, Stem Cell Markers | |
| 1280 | |
| Human, Mouse, Rat, Chicken, Rabbit | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu