Learn More
CLIP Mouse anti-Human, Brilliant Violet 605, Clone: CerCLIP, BD Optibuild™
Mouse Monoclonal Antibody
Marque: BD Biosciences 742597
Description
The CerCLIP monoclonal antibody specifically recognizes the class II associated invariant chain peptides (CLIP). CLIP is the remaining segment of class II invariant chain (Ii) after proteolytic degradation which stays associated with HLA-DR and represents a final intermediate in the removal of invariant chain from class II molecules during antigen processing. The removal of CLIP and subsequent loading of the antigenic peptide is facilitated by the non-classical class II molecule HLA-DM. CLIP can be detected, in association with HLA-DR, on the surface of T2DR3, a mutant cell line lacking HLA-DM.
This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
Spécification
| CLIP | |
| CerCLIP | |
| Brilliant Violet 605 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.