Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, Natural Kynar™
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Marque:  Cole-Parmer 50109-20
Code produit. 30053467
  • 36.50€ / 10 pièces
Expédition estimée: 21-07-2025
pour voir le stock disponible 
Découvrez plus d'offres spéciales
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Spécification
Spécification
Non-DEHP;Non-Phthalate;USP
10/Pk.
Hose Barb Reducer
Structuré
-35°C
1/2 to 1/4 in.
E-beam/gamma irradiation, EtO, or autoclave up to 135°C
Naturel
Nom du produit
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, Natural Kynar™ > 1/2 to 1/4 in.