missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CISD1 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16006-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognizing MitoNEET (calculated 12 kDa) as a 17 kDa protein may be due to its posttranslational modification or metal binding activity.
This gene encodes a protein with a CDGSH iron-sulfur domain and has been shown to bind a redox-active [2Fe-2S] cluster. The encoded protein has been localized to the outer membrane of mitochondria and is thought to play a role in regulation of oxidation. Genes encoding similar proteins are located on chromosomes 4 and 17, and a pseudogene of this gene is located on chromosome 2.Specifications
CISD1 | |
Polyclonal | |
Unconjugated | |
CISD1 | |
C10orf70, CDGSH iron sulfur domain 1, CISD1, MDS029, mitoNEET, Mnt, ZCD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
294362, 393577, 52637, 55847 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B0K020, Q91WS0, Q9NZ45 | |
Cisd1 | |
CISD1 Fusion Protein Ag8680 | |
150 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |