missing translation for 'onlineSavingsMsg'
Learn More

CHSY1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marque:  Invitrogen PA550008

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.01000kg

Code produit. 15693935

  • 472.00€ / 100µL
Expédition estimée: 20-09-2024
pour voir le stock disponible 

Description

Description

The antibody detects endogenous levels of total CHSY1 protein.

CHSY1 synthesizes chondroitin sulfate, a glycosaminoglycan expressed on the surface of most cells and in extracellular matrices. Glycosaminoglycan chains are covalently linked to a wide range of core protein families and regulate many biologic processes, including cell proliferation and recognition, extracellular matrix deposition, and morphogenesis.
Spécification

Spécification

CHSY1
Polyclonal
Unconjugated
CHSY1
carbohydrate (chondroitin) synthase 1; carbohydrate synthase 1; chondroitin glucuronyltransferase 1; chondroitin glucuronyltransferase II; chondroitin sulfate synthase 1; Chondroitin synthase 1; CHSY; CHSY1; ChSy-1; chys1; CSS1; glucuronosyl-N-acetylgalactos; glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase 1; KIAA0990; mKIAA0990; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase 1; N-acetylgalactosaminyltransferase 1; N-acetylgalactosaminyltransferase II; TPBS; UNQ756/PRO1487; wu:fc27h05; wu:fc27h06; zgc:64191
Rabbit
Antigen affinity chromatography
RUO
22856
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q86X52
CHSY1
Synthesized peptide derived from internal of human CHSY1.
100 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions