missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ChREBP Polyclonal Antibody, DyLight™ 650, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA522922
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
The target sequence has 83% sequence homology with mouse ChREBP.
ChREBP is a transcription factor involved in activating genes that encode enzymes of fatty acid biosynthesis in liver and adipose. This protein may have relevance to lipid metabolism, obesity, and type II diabetes.Spécification
ChREBP | |
Polyclonal | |
DyLight 650 | |
MLXIPL | |
BHLHD14; carbohydrate response element binding protein; carbohydrate-responsive element-binding protein; ChREBP; Class D basic helix-loop-helix protein 14; MIO; MLX interacting protein like; MLX interacting protein-like; MLX interacting protein-like beta; Mlx interactor; MLX-interacting protein-like; MLXIPL; MONDOB; putative hepatic transcription factor; WBSCR14; Williams Beuren syndrome chromosome region 14; williams-Beuren syndrome chromosomal region 14 protein; Williams-Beuren syndrome chromosomal region 14 protein homolog; Williams-Beuren syndrome chromosome region 14 homolog; Williams-Beuren syndrome chromosome region 14 protein 1; Williams-Beuren syndrome chromosome region 14 protein 2; Williams-Beuren syndrome chromosome region 14 protein 3; WS basic-helix-loop-helix leucine zipper protein; WS-bHLH | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
171078, 51085, 58805 | |
4° C, store in dark | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
50mM sodium borate with no preservative | |
Q8VIP2, Q99MZ3, Q9NP71 | |
MLXIPL | |
An C-terminal synthetic peptide made to the human ChREBP protein sequence (between residues 800-852). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |