missing translation for 'onlineSavingsMsg'
Learn More

Chk1 Mouse anti-Human, Mouse, Rat, Clone: 3A3A5, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 60277-1-IG-150UL

 Afficher plus de versions de ce produit

Code produit. 16897335

  • 410.00€ / 150µL

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Découvrez plus d'offres spéciales
This item is not returnable. View return policy

Description

Description

CHK1 (CHEK1) is a kinase that phosphorylates cdc25, an important phosphatase in cell cycle control, particularly for entry into mitosis. CHK1 can also phosphorylate p53 at Ser20 in vitro. Cell cycle events are regulated by the sequential activation and deactivation of cyclin dependent kinases (Cdks) and by proteolysis of cyclins. CHK1 is involved in these processes as regulators of Cdks. CHK1 function as an essential component in the G2 DNA damage checkpoint by phosphorylating Cdc25C in response to DNA damage. Phosphorylation inhibits Cdc25C activity, thereby blocking mitosis. Cdc25A, Cdc25B and Cdc25C protein tyrosine phosphatases function as mitotic activators by dephosphorylating Cdc2 p34 on regulatory tyrosine residues. CHK1 can phosphorylate Wee 1 in vitro, providing evidence that the hyperphosphorylated form of Wee 1, seen in cells delayed by CHK1 overexpression, is due to phosphorylation by CHK1. CHK1 is phosphorylated on Serine 345 (S345) in response to UV, IR and hydroxyurea (HU). CHK1 plays an essential role in the mammalian DNA damage checkpoint, embryonic development and tumor suppression. Further, CHK1 is a key mediator in the DNA damage-induced checkpoint network. CHK1 is an evolutionarily conserved protein kinase that functions to ensure genomic integrity upon genotoxic stress. When the G2 or S checkpoint is abrogated by the inhibition of CHK1, p53-deficient cancer cells undergo mitotic catastrophe and eventually apoptosis, whereas normal cells are still arrested in the G1 phase. Thus, CHK1 inhibitors can preferentially potentiate the efficacy of DNA damaging agents in cancer cells, and Chk1 is an attractive therapeutic target for cancer treatment, especially since approximately 50% of all human cancers are p53-deficient.
Specifications

Specifications

Chk1
Monoclonal
1.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14757, O35280, Q91ZN7
Chek1
Chk1 Fusion Protein Ag0409
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
3A3A5
Unconjugated
CHEK1
CHEK1, Chk1
Mouse
Protein A
RUO
1111, 12649, 140583
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers