missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGS 19755, Tocris Bioscience™
Click to view available options
Quantité:
10 mg
50 mg
Description
Potent, competitive NMDA receptor antagonist. Anticonvulsant and neuroprotective.
Spécification
Spécification
Nom chimique ou matériau | CGS 19755 |
Quantité | 50 mg |
Poids de la formule | 223.17 |
Pourcentage de pureté | >98% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'