missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK2, p Thr160 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
196.00€ - 466.00€
Spécification
Antigène | CDK2 |
---|---|
Dilution | Western Blot 1:100 - 1:500, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18602407
|
Novus Biologicals
NBP3-05409-100ul |
100 μg |
466.00€
100µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
18642429
|
Novus Biologicals
NBP3-05409-20ul |
20 μg |
196.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
CDK2 Polyclonal antibody specifically detects CDK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Spécification
CDK2 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Cell Cycle and Replication, Ovarian Carcinoma Cell Markers, Phospho Specific | |
PBS with 50% glycerol, pH7.3. | |
1017 | |
IgG | |
Affinity purified |
Western Blot 1:100 - 1:500, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
cdc2-related protein kinase, cell devision kinase 2, Cell division protein kinase 2, cyclin-dependent kinase 2, EC 2.7.11, EC 2.7.11.22, p33 protein kinase, p33(CDK2) | |
A synthetic phosphorylated peptide around T160 of human CDK2 (NP_001789.2?/NP_001249.1). TYTHE | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |