Learn More
CD8 Mouse anti-Human, CoraLite 488, Clone: SK1, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech CL488-65146-100TESTS
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CD8 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD8. This antibody recognizes human antigen. The CD8 antibody has been validated for the following applications: FC analysis. This antibody is coralite™488.
Spécification
CD8 | |
Monoclonal | |
5 μL/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
SK1 | |
CoraLite 488 | |
100 Tests | |
925 | |
IgG1 κ |