Learn More
CD6 Monoclonal Antibody (MEM-98), PE, Invitrogen™
Mouse Monoclonal Antibody
Marque:  Invitrogen MA110258
Code produit. 12362273
  • 473.00€ / 100 pièces
Découvrez plus d'offres spéciales
Description
Description
CD6, also known as T12, is a member of the scavenger receptor superfamily found on T and B cell subsets, thymocytes, and acute lymphocytic leukemia cells (ALL). CD6 interacts with its ligand CD166/ALCAM (activated leukocyte cell adhesion molecule) and serves as a coreceptor for T cell activation and stabilizer of the immunological synapse. CD6-ALCAM mediated cell adhesion is also important for T cell proliferation. CD6 may exert some its functions via association with CD5, probably by fine-tuning CD5 signaling. Ligation of CD6 has antiapoptotic role in chronic lymphocytic leukemia B cells.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
CD6
Monoclonal
PE
CD6
Cd6; CD6 antigen; CD6 antigen, Tp120; Cd6 molecule; FLJ44171; OX52; Soluble CD6; T12; T-cell differentiation antigen CD6; TP120
Mouse
Size-exclusion chromatography
RUO
923
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
MEM-98
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P30203
CD6
Human CD6 antigen purified by immunoaffinity chromatography from HBP-ALL cells followed by preparative SDS-PAGE of non-boiled non-reduced sample (excised piece of gel corresponding to the 100 kDa zone).
100 Tests
Primary
Human
Antibody
IgG1