missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD56 Monoclonal Antibody (MEM-188), PerCP

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA110255
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody reacts with an extracellular epitope on a 180 kDa isoform of CD56 (NCAM) expressed in leukocytes. It has been suggested that the antibody MEM-188 could react with rhesus monkey lymphocytes. Reactivity with other NCAM isoforms has not been tested.
CD56, also known as neural cell adhesion molecule (NCAM), is a highly glycosylated transmembrane glycoprotein of the immunoglobulin family. It plays a crucial role in cell adhesion, migration, axonal growth, pathfinding, and synaptic plasticity. CD56 is ubiquitously expressed in the nervous system in isoforms ranging from 120-180 kDa and is involved in homotypic adhesion of neural cells. It mediates interactions by binding extracellular matrix components such as laminin and integrins, with polysialic modification reducing CD56-mediated adhesion. In the hematopoietic system, CD56 is expressed on natural killer (NK) cells and a subset of T cells known as NKT cells. It is also found on most neuroectodermal-derived cell lines, tissues, and neoplasms, including retinoblastoma, medulloblastoma, astrocytomas, and neuroblastoma. CD56 serves as a widely used neuroendocrine marker with high sensitivity for neuroendocrine tumors and ovarian granulosa cell tumors. Diseases associated with CD56 dysfunction include rabies and blastic plasmacytoid dendritic cell neoplasms, highlighting its importance in both neural and immune system functions.
Spécification
| CD56 | |
| Monoclonal | |
| 0.07 mg/mL | |
| PBS with 0.2% BSA and 15mM sodium azide | |
| P13591 | |
| Ncam1 | |
| KG-1 human acute myelogenous leukemia cell line. | |
| 100 Tests | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| MEM-188 | |
| PerCP | |
| Ncam1 | |
| adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| 4684 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu