missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD46 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12494-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD46 (MCP, membrane cofactor protein) is a multifunctional cell surface transmembrane protein that binds and inactivates C3b and C4b complement fragments, regulates T cell-induced inflammatory responses by either inhibiting (CD46-1 isoform) or increasing (CD46-2 isoform) the contact hypersensitivity reaction. CD46 also serves as a receptor for several human pathogens (both bacteria and viruses), and its ligation alteres T lymphocyte polarization toward antigen-presenting cells or target cells, inhibiting lymphocyte function. CD46 is a protector of placental tissue and is also expressed on the inner acrosomal membrane of spermatozoa.Spécification
CD46 | |
Polyclonal | |
Unconjugated | |
CD46 | |
CD46, MCP, Membrane cofactor protein, MIC10, TLX, TRA2.10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17221, 4179 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88174, P15529 | |
Cd46 | |
CD46 Fusion Protein Ag3131 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |