Learn More
CD45RA Mouse anti-Human, CoraLite 647, Clone: HI100, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech CL647-65108-100TESTS
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CD45RA antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD45RA. This antibody recognizes human antigen. The CD45RA antibody has been validated for the following applications: FC analysis. This antibody is coralite™647.
Spécification
CD45RA | |
Monoclonal | |
5 μL/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
HI100 | |
CoraLite 647 | |
100 Tests | |
5788 | |
IgG2b |