Learn More
CD45 Mouse, PerCP, Clone: 2D1, enQuireBio™
Mouse Monoclonal Antibody
Marque:  EnquireBio QAB97-PCP-25Tests
Code produit. 15972648
  • 149.00€ / 25 tests
Découvrez plus d'offres spéciales
Spécification
CD45
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
PTPRC
Protein A/G Purified
RUO
5788
IgG1 κ
Flow Cytometry
2D1
PerCP
NP_002829
Mouse
25 Tests
Primary
Store at 2-4°C.